Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00628199_m1
          See other ARFGAP1 GE Assays ›
          Gene Symbol:
          Arfgap1
          Gene Name:
          ADP-ribosylation factor GTPase activating protein 1
          Gene Aliases:
          AI115377, Arf1gap
          Chromosome Location:
          Chr.2: 180967225 - 180982524 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00628199_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Arfgap1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001177706.1 NP_001171177.1 5 - 6 957 92
          NM_001177707.1 NP_001171178.1 6 - 7 828 92
          NM_001177708.1 NP_001171179.1 5 - 6 547 92
          NM_001177709.1 NP_001171180.1 5 - 6 547 92
          NM_001177710.1 NP_001171181.1 4 - 5 437 92
          NM_145760.3 NP_665703.2 6 - 7 760 92
          GenBank mRNA AK030295.1 - 6 - 7 716 92
          AK030520.1 - 5 - 6 959 92
          AK134402.1 - 6 - 7 770 92
          AK145879.1 - 6 - 7 744 92
          AK168007.1 - 6 - 7 741 92
          AK168602.1 - 5 - 6 528 92
          BC016228.1 - 4 - 5 395 92
          BC052922.1 - 5 - 6 495 6312351 92
          BC059817.1 - 5 - 6 486 5717009 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002811.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Arfgap1

          Panther Classification:

          Molecular Function -

          G-protein modulator protein-binding activity modulator GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          protein transport
          vesicle-mediated transport
          regulation of endocytosis
          positive regulation of GTPase activity
          GTPase activator activity
          protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_001177710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_145760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 NM_145760.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023036.mm.1 TC0200002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_001177710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_145760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 NM_145760.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023033.mm.1 TC0200002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_001177710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_145760.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 NM_145760.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200023031.mm.1 TC0200002811.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline