Search
Search

Vps36| Gene Symbol: Vps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027338.1 | NP_081614.1 | 7 - 8 | 574 | 97 | |
| GenBank mRNA | AK150754.1 | - | 7 - 8 | 595 | 97 | |
| AK151035.1 | - | 7 - 8 | 595 | 97 | ||
| AK161865.1 | - | 626 | 97 | |||
| BC010811.1 | - | 7 - 8 | 574 | 4210911 | 97 | |
Set Membership: |
|