Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00777741_sH
          See other MRPL32 GE Assays ›
          Gene Symbol:
          Mrpl32
          Gene Name:
          mitochondrial ribosomal protein L32
          Gene Aliases:
          0610033O15Rik
          Chromosome Location:
          Chr.13: 14610301 - 14613037 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00777741_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl32
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029271.2 NP_083547.1 2 247 110
          GenBank mRNA AB049650.1 - 2 278 110
          AF279255.1 - 2 224 110
          AK002753.1 - 2 248 110
          AK031791.1 - 1 1524 110
          AK078324.1 - 2 265 110
          BC052214.1 - 2 282 30018135 110

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl32

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839394 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839397 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839395 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 AK031791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 AK078324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 BC037368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 BC052214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 NM_029271.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 AK031791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 AK078324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 BC037368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 BC052214.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P141818 NM_029271.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 596104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 97503 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126541 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 AK031791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 AK078324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 BC037368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 BC052214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 NM_029271.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 AK031791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 AK078324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 BC037368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 BC052214.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P141818 NM_029271.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 AK031791.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 AK078324.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 BC037368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 BC052214.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 NM_029271.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 AK031791.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 AK078324.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 BC037368.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 BC052214.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P141818 NM_029271.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839380 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839397 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839361 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839366 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28839382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126541 - -
          Whole Mouse Genome G4122A A_51_P141818 AK031791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 AK078324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 BC037368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 BC052214.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 NM_029271.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 AK031791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 AK078324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 BC037368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 BC052214.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P141818 NM_029271.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105376 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126541 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10786995 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10523767 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10786996 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10786985 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10523769 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839397 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839396 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839395 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28839394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28839395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10948519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839355 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10905189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839393 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10791330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839350 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10905185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11029124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839346 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28839348 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10407872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407875 - -
          Affymetrix HT MG-430 PM Array Plate 1424372_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10407870 - -
          Mouse Genome 430 2.0 Array 1424372_PM_at - -
          Mouse Genome 430 2.0 Array 1424372_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75398_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75398_at - -
          Affymetrix Mouse Clariom S Array PSR1300009450.mm.1 NM_029271.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009450.mm.1 NM_029271.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009450.mm.1 TC1300001479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009453.mm.1 TC1300001479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009445.mm.1 NM_029271.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009445.mm.1 NM_029271.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009445.mm.1 TC1300001479.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300009447.mm.1 NM_029271.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009447.mm.1 NM_029271.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300009447.mm.1 TC1300001479.mm.1 MTA TC
          Mouse Expression Set 430 1424372_at - -
          Mouse Expression Set 430 1424372_PM_at - -
          Mouse Genome 430A 2.0 Array 1424372_PM_at - -
          Mouse Genome 430A 2.0 Array 1424372_at - -
          Affymetrix HT Mouse Genome 430A Array 1424372_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424372_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 640249 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 110041 - -
          Illumina mouse-6 v1.0 expression beadchip 106660706 - -
          Illumina mouse-6 v1.0 expression beadchip 110041 - -
          Illumina mouse-6 v1.0 expression beadchip 100780348 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110041 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2695342 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2431602 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2427238 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2695342 - -
          mouse-6 v1.1 (Illumina) scl0075398.1_198-S - -
          mouse-6 v1.1 (Illumina) scl44271.1_83-S - -
          mouse-6 v1.1 (Illumina) scl0003660.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 110041 - -
          Illumina Sentrix mouse-6.v1 bead array 106660706 - -
          Illumina Sentrix mouse-6.v1 bead array 100780348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0075398.1_198-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100780348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110041 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106660706 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2695342 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2695342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline