Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00844325_s1
          See other 2610305D13RIK GE Assays ›
          Gene Symbol:
          2610305D13Rik
          Gene Name:
          RIKEN cDNA 2610305D13 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 147611936 - 147642508 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00844325_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2610305D13Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145078.2 NP_659544.2 5 1655 93
          GenBank mRNA AK011986.1 - 2 1357 93
          BC032186.1 - 1 1143 93
          BC058858.1 - 5 1659 93
          BC138179.1 - 2 1298 8861198 93
          BC145854.1 - 2 1298 40131157 93

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2610305D13Rik

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037050 NM_001177767.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037050 NM_001103158.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037050 BC096593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037050 NM_001177545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037050 NM_001177546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037081 DV659285.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037081 BC032186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037081 BC058858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2037081 NM_145078.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 119647 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037050 NM_001177767.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037050 NM_001103158.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037050 BC096593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037050 NM_001177545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037050 NM_001177546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037081 DV659285.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037081 BC032186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037081 BC058858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2037081 NM_145078.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347875 DV659285.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347875 AK011986.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347875 BC032186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347875 BC058858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P347875 NM_145078.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23390010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23390004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23390006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23390002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23390009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23390000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23389975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04048533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390002 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390009 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04048529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03658581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03658584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23390004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23389999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03658567 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518360 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518359 - -
          Affymetrix HT MG-430 PM Array Plate 1428604_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518358 - -
          Mouse Genome 430 2.0 Array 1428604_PM_at - -
          Mouse Genome 430 2.0 Array 1428604_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 112422_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 112422_at - -
          Affymetrix Mouse Clariom S Array PSR0400032942.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032942.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032944.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032944.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032951.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032951.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032943.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032943.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032940.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032940.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032941.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032941.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032945.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032945.mm.1 NM_145078.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032949.mm.1 TC0400003992.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032949.mm.1 NM_145078.2 Refseq
          Mouse Expression Set 430 1428604_PM_at - -
          Mouse Expression Set 430 1428604_at - -
          Murine Genome U74v2 Set 165755_r_at - -
          Murine Genome U74v2 Set 165755_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1428604_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428604_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 359618 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360603 - -
          Illumina mouse-6 v1.0 expression beadchip 3360603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2776728 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2776728 - -
          mouse-6 v1.1 (Illumina) scl23536.7_579-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23536.7_579-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2998406 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2776728 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2998406 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline