Search
Search

Polrmt| Gene Symbol: Polrmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172551.3 | NP_766139.2 | 14 - 15 | 3094 | 81 | |
| GenBank mRNA | AK003792.2 | - | 5 - 6 | 832 | 81 | |
| AK030039.1 | - | 13 - 14 | 2876 | 81 | ||
| AK042071.1 | - | 12 - 13 | 2651 | 81 | ||
| AK047545.1 | - | 12 - 13 | 3435 | 81 | ||
| AK053356.1 | - | 14 - 15 | 3085 | 81 | ||
| AK085666.1 | - | 14 - 15 | 3087 | 81 | ||
| AK128956.1 | - | 14 - 15 | 2787 | 81 | ||
| AK154729.1 | - | 14 - 15 | 3076 | 81 | ||
| BC110697.1 | - | 14 - 15 | 3070 | 30356222 | 81 | |
| BC117939.1 | - | 14 - 15 | 3087 | 81 | ||
| BC117940.1 | - | 14 - 15 | 3087 | 40097816 | 81 | |
Set Membership: |
|