Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01151455_g1
          See other COMMD4 GE Assays ›
          Gene Symbol:
          Commd4
          Gene Name:
          COMM domain containing 4
          Gene Aliases:
          1110039H05Rik
          Chromosome Location:
          Chr.9: 57155035 - 57158349 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01151455_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Commd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025417.2 NP_079693.1 7 - 8 584 59
          GenBank mRNA AK004174.1 - 7 - 8 585 59
          AK009802.1 - 7 - 8 583 59
          AK172194.1 - 6 - 7 1696 59
          BC033438.1 - 6 - 7 570 3995599 59
          BC049222.1 - 7 - 8 583 2649359 59
          BC055798.1 - 6 - 7 1675 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002366.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Commd4

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 BC055798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 AK172194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 BC033438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 BC049222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 NM_025417.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 BC055798.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 AK172194.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 BC033438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 BC049222.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353592 NM_025417.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929490 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 184754 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 BC055798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 AK172194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 BC033438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 BC049222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 NM_025417.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 BC055798.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 AK172194.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 BC033438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 BC049222.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353592 NM_025417.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 BC055798.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 AK172194.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 BC033438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 BC049222.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 NM_025417.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 BC055798.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 AK172194.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 BC033438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 BC049222.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P353592 NM_025417.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26540608 - -
          Whole Mouse Genome G4122A A_51_P353592 BC055798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 AK172194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 BC033438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 BC049222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 NM_025417.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 BC055798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 AK172194.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 BC033438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 BC049222.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353592 NM_025417.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07839216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07740295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07839213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07839205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26540631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07625248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540608 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07740295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07640713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07625247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07640710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07640725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540602 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26540623 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10593904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593910 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593905 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593909 - -
          Affymetrix HT MG-430 PM Array Plate 1417006_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593903 - -
          Mouse Genome 430 2.0 Array 1417006_PM_at - -
          Mouse Genome 430 2.0 Array 1417006_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66199_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66199_at - -
          Affymetrix Mouse Clariom S Array PSR0900019808.mm.1 NM_025417.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019808.mm.1 NM_025417.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900019808.mm.1 TC0900002366.mm.1 MTA TC
          Mouse Expression Set 430 1417006_PM_at - -
          Mouse Expression Set 430 1417006_at - -
          Murine Genome U74v2 Set 99167_at - -
          Murine Genome U74v2 Set 99167_PM_at - -
          Murine Genome U74v2 Set 170231_PM_i_at - -
          Murine Genome U74v2 Set 169728_PM_at - -
          Murine Genome U74v2 Set 170470_r_at - -
          Murine Genome U74v2 Set 170231_i_at - -
          Murine Genome U74v2 Set 170470_PM_r_at - -
          Murine Genome U74v2 Set 169728_at - -
          Mouse Genome 430A 2.0 Array 1417006_at - -
          Mouse Genome 430A 2.0 Array 1417006_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417006_at - -
          Affymetrix HT Mouse Genome 430A Array 1417006_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 510110 - -
          Illumina mouse-6 v1.0 expression beadchip 510110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213206 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213206 - -
          mouse-6 v1.1 (Illumina) scl35797.7.1_123-S - -
          Illumina Sentrix mouse-6.v1 bead array 510110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35797.7.1_123-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2798183 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2798183 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35797.7.1_123-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline