Search
Search

Slc25a13| Gene Symbol: Slc25a13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177572.1 | NP_001171043.1 | 8 - 9 | 1010 | 81 | |
| NM_015829.3 | NP_056644.1 | 8 - 9 | 1010 | 81 | ||
| GenBank mRNA | AF164632.1 | - | 8 - 9 | 955 | 81 | |
| AK002829.1 | - | 8 - 9 | 885 | 81 | ||
| AK012670.1 | - | 8 - 9 | 901 | 81 | ||
| AK029020.1 | - | 8 - 9 | 941 | 81 | ||
| AK150496.1 | - | 8 - 9 | 956 | 81 | ||
| AK151180.1 | - | 8 - 9 | 956 | 81 | ||
| AK152237.1 | - | 8 - 9 | 956 | 81 | ||
| BC016571.1 | - | 2 - 3 | 147 | 3600909 | 81 | |
Set Membership: |
|