Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01165164_m1
          See other OXR1 GE Assays ›
          Gene Symbol:
          Oxr1
          Gene Name:
          oxidation resistance 1
          Gene Aliases:
          2210416C20Rik, C7, C7B
          Chromosome Location:
          Chr.15: 41447482 - 41861048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01165164_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Oxr1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001130163.1 NP_001123635.1 8 - 9 1655 107
          NM_001130165.1 NP_001123637.1 8 - 9 1750 107
          NM_001130166.1 NP_001123638.1 10 - 11 2014 107
          NM_130885.2 NP_570955.1 8 - 9 1655 107
          GenBank mRNA AF324899.1 - 8 - 9 1654 107
          AF333985.1 - 8 - 9 1655 107
          AK040881.1 - 8 - 9 1678 107
          AK158703.1 - 10 - 11 1996 107
          BC003927.1 - 2 - 3 245 107
          BC089183.1 - 8 - 9 1745 5321255 107
          BC098491.1 - 10 - 11 1997 30086857 107

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the BC003927.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Oxr1

          Gene Ontology Categories:

          Function(s) Process(es)

          adult walking behavior
          negative regulation of neuron apoptotic process
          oxidation-reduction process
          cellular response to hydroperoxide
          oxidoreductase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_001130163.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_001130163.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_001130165.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_001130165.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_001130166.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_001130166.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_130885.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 NM_130885.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002328.mm.1 TC1500000331.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_001130163.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_001130163.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_001130165.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_001130165.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_001130166.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_001130166.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_130885.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 NM_130885.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500002327.mm.1 TC1500000331.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline