Search
Search

Smpd4| Gene Symbol: Smpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164609.1 | NP_001158081.1 | 9 - 10 | 916 | 79 | |
| NM_001164610.1 | NP_001158082.1 | 9 - 10 | 916 | 79 | ||
| NM_001164611.1 | NP_001158083.1 | 9 - 10 | 916 | 79 | ||
| NM_029945.3 | NP_084221.2 | 9 - 10 | 916 | 79 | ||
| GenBank mRNA | AK019466.2 | - | 9 - 10 | 917 | 79 | |
| AK031029.1 | - | 9 - 10 | 910 | 79 | ||
| AK031685.1 | - | 9 - 10 | 915 | 79 | ||
| AK035892.1 | - | 9 - 10 | 908 | 79 | ||
| AK036055.1 | - | 9 - 10 | 908 | 79 | ||
| AK036380.1 | - | 9 - 10 | 912 | 79 | ||
| AK129354.1 | - | 9 - 10 | 893 | 79 | ||
| AK138720.1 | - | 9 - 10 | 890 | 79 | ||
| AK164234.1 | - | 9 - 10 | 908 | 79 | ||
| AK164456.1 | - | 7 - 8 | 1381 | 79 | ||
Set Membership: |
|