Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01171299_m1
          See other HIPK4 GE Assays ›
          Gene Symbol:
          Hipk4
          Gene Name:
          homeodomain interacting protein kinase 4
          Gene Aliases:
          Gm162
          Chromosome Location:
          Chr.7: 27523261 - 27531180 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01171299_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hipk4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033315.2 NP_001028487.2 3 - 4 1930 72
          GenBank mRNA AB354583.1 - 3 - 4 1674 72
          AK133498.1 - 2 - 3 2447 72
          BC147572.1 - 2 - 3 1090 9056562 72
          BC147577.1 - 2 - 3 1090 9056567 72
          CB248531.1 - 1 - 2 474 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000462.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          histone phosphorylation
          peptidyl-serine phosphorylation
          protein autophosphorylation
          regulation of signal transduction by p53 class mediator
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045414 BC147577.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045414 CJ235960.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045414 BC147572.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2045414 NM_001033315.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 676314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193247 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407080 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 135059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944431 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 74897 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172511 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045414 BC147577.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045414 CJ235960.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045414 BC147572.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2045414 NM_001033315.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172511 - -
          Whole Mouse Genome G4122A A_52_P124526 BC147577.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P124526 CJ235960.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P124526 AK133498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P124526 BC147572.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P124526 NM_001033315.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25017713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017695 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06516121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06000212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06631587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06631596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06631589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06631588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06335683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06497268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06578891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06260960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06516120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06000215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06578594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25017719 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551381 - -
          Affymetrix Mouse Gene 1.1 ST Array 10551377 - -
          Affymetrix Mouse Clariom S Array PSR0700003681.mm.1 NM_001033315.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003681.mm.1 NM_001033315.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003681.mm.1 TC0700000462.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003680.mm.1 NM_001033315.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003680.mm.1 NM_001033315.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003680.mm.1 TC0700000462.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 300400 - -
          ABI Mouse Genome Survey Microarray 818809 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244376 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2794023 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2794023 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline