Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01173994_m1
          See other LYSMD2 GE Assays ›
          Gene Symbol:
          Lysmd2
          Gene Name:
          LysM, putative peptidoglycan-binding, domain containing 2
          Gene Aliases:
          2210402C18Rik, AV033872, AW538442
          Chromosome Location:
          Chr.9: 75625732 - 75637773 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01173994_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lysmd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027309.2 NP_081585.2 2 - 3 634 58
          GenBank mRNA BC048545.1 - 2 - 3 623 6589154 58
          DT905233.1 - 2 - 3 389 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001018.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lysmd2

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645025 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645017 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645018 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645020 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233160 NM_027309.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233160 AK008800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233160 BC048545.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233160 NM_027309.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233160 AK008800.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P233160 BC048545.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 58036 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 591577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893005 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 384875 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139316 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148308 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P122260 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233160 NM_027309.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233160 AK008800.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233160 BC048545.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233160 NM_027309.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233160 AK008800.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P233160 BC048545.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645005 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26645028 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01974743 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148308 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P122260 - -
          Whole Mouse Genome G4122A A_51_P233160 NM_027309.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233160 AK008800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233160 BC048545.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233160 NM_027309.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233160 AK008800.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P233160 BC048545.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129915 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148308 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P122260 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07907937 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07773987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07800891 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07907936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07576155 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07576160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07576157 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645020 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645024 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645017 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645018 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645023 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26645025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26644993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26644995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26644994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26645055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07987200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07517903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07576157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08054135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08105131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08054129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07854919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26644998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07907944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07773985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26645013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08009185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10587230 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587229 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587228 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587227 - -
          Affymetrix HT MG-430 PM Array Plate 1447536_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428626_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10587226 - -
          Mouse Genome 430 2.0 Array 1447536_PM_at - -
          Mouse Genome 430 2.0 Array 1428626_PM_at - -
          Mouse Genome 430 2.0 Array 1428626_at - -
          Mouse Genome 430 2.0 Array 1447536_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70082_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70082_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900008052.mm.1 NM_027309.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008052.mm.1 NM_027309.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008052.mm.1 TC0900001018.mm.1 MTA TC
          Mouse Expression Set 430 1428626_PM_at - -
          Mouse Expression Set 430 1428626_at - -
          Mouse Expression Set 430 1447536_at - -
          Mouse Expression Set 430 1447536_PM_at - -
          Murine Genome U74v2 Set 103560_PM_at - -
          Murine Genome U74v2 Set 103560_at - -
          Mouse Genome 430A 2.0 Array 1428626_PM_at - -
          Mouse Genome 430A 2.0 Array 1428626_at - -
          Affymetrix HT Mouse Genome 430A Array 1428626_at - -
          Affymetrix HT Mouse Genome 430A Array 1428626_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447536_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1447536_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 668792 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2970037 - -
          Illumina mouse-6 v1.0 expression beadchip 2970037 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002970037 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239808 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239808 - -
          mouse-6 v1.1 (Illumina) scl0070082.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 2970037 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0070082.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2970037 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1239808 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239808 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0070082.1_20-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline