Search
Search

Hmg20b| Gene Symbol: Hmg20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163165.1 | NP_001156637.1 | 7 - 8 | 765 | 207 | |
| NM_001163166.1 | NP_001156638.1 | 7 - 8 | 662 | 207 | ||
| NM_010440.3 | NP_034570.1 | 6 - 7 | 849 | 207 | ||
| GenBank mRNA | AF146224.1 | - | 6 - 7 | 832 | 207 | |
| AK153816.1 | - | 7 - 8 | 663 | 207 | ||
| AK155652.1 | - | 7 - 8 | 667 | 207 | ||
| AK155890.1 | - | 6 - 7 | 725 | 207 | ||
| AL355735.1 | - | 3 - 4 | 224 | 207 | ||
| BC003924.1 | - | 6 - 7 | 737 | 3485302 | 207 | |
| BC011334.1 | - | 7 - 8 | 698 | 4012515 | 207 | |
| BC023436.1 | - | 4 - 5 | 325 | 207 | ||
| BC071217.1 | - | 4 - 5 | 1726 | 207 | ||
Set Membership: |
|