Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184918_g1
          See other WDR54 GE Assays ›
          Gene Symbol:
          Wdr54
          Gene Name:
          WD repeat domain 54
          Gene Aliases:
          1700030E05Rik
          Chromosome Location:
          Chr.6: 83152710 - 83156406 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184918_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wdr54
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023790.2 NP_076279.1 9 - 10 1076 68
          GenBank mRNA AF053628.1 - 9 - 10 1064 68
          AK006536.1 - 9 - 10 976 68
          AK154345.1 - 9 - 10 1067 68
          BC083332.1 - 10 - 11 981 6819838 68
          BY540708.1 - 2 - 3 179 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002562.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Wdr54

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591794 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591796 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 AK154345.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 BC083332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 AK006536.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 NM_023790.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 AF053628.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 AK154345.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 BC083332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 AK006536.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 NM_023790.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P424722 AF053628.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 632408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802348 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156309 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 AK154345.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 BC083332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 AK006536.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 NM_023790.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 AF053628.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 AK154345.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 BC083332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 AK006536.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 NM_023790.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P424722 AF053628.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591773 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591777 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24591755 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156309 - -
          Whole Mouse Genome G4122A A_51_P424722 AK154345.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 BC083332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 AK006536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 NM_023790.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 AF053628.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 AK154345.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 BC083332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 AK006536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 NM_023790.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P424722 AF053628.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140590 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05785195 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05785194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05785191 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591793 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591792 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591794 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591797 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591799 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591771 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591772 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591798 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591796 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591773 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24591814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24591682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05032533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05440010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591813 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05785194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05032542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05455060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05032535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05250857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05710736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05250849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05785185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05785191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591781 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05032541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24591752 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10545665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545671 - -
          Affymetrix Mouse Gene 1.0 ST Array 10545663 - -
          Affymetrix HT MG-430 PM Array Plate 1422616_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1425980_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10545658 - -
          Mouse Genome 430 2.0 Array 1422616_s_at - -
          Mouse Genome 430 2.0 Array 1422616_PM_s_at - -
          Mouse Genome 430 2.0 Array 1425980_PM_at - -
          Mouse Genome 430 2.0 Array 1425980_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75659_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75659_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600019864.mm.1 NM_023790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019864.mm.1 NM_023790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600019864.mm.1 TC0600002562.mm.1 MTA TC
          Mouse Expression Set 430 1425980_at - -
          Mouse Expression Set 430 1422616_PM_s_at - -
          Mouse Expression Set 430 1425980_PM_at - -
          Mouse Expression Set 430 1422616_s_at - -
          Murine Genome U74v2 Set 167048_at - -
          Murine Genome U74v2 Set 167048_PM_at - -
          Mouse Genome 430A 2.0 Array 1422616_s_at - -
          Mouse Genome 430A 2.0 Array 1422616_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1425980_at - -
          Mouse Genome 430A 2.0 Array 1425980_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422616_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1425980_at - -
          Affymetrix HT Mouse Genome 430A Array 1422616_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1425980_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 399526 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1740433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2755692 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2755692 - -
          mouse-6 v1.1 (Illumina) scl075659.3_69-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl075659.3_69-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2755692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2755692 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline