Search
Search
| Gene Symbol: Pet2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008821.2 | NP_032847.2 | 6 - 7 | 737 | 62 | |
| GenBank mRNA | BC138080.1 | - | 5 - 6 | 541 | 8861099 | 62 |
| M95564.1 | - | 6 - 7 | 739 | 62 | ||
Set Membership: |
|