Search
Search

Tnfaip3| Gene Symbol: Tnfaip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166402.1 | NP_001159874.1 | 6 - 7 | 2049 | 104 | |
| NM_009397.3 | NP_033423.3 | 7 - 8 | 2134 | 104 | ||
| GenBank mRNA | AK088297.1 | - | 7 - 8 | 2091 | 104 | |
| AK151921.1 | - | 7 - 8 | 2095 | 104 | ||
| AK153238.1 | - | 7 - 8 | 2089 | 104 | ||
| AK172529.1 | - | 6 - 7 | 2051 | 104 | ||
| BC054806.1 | - | 7 - 8 | 2134 | 6838303 | 104 | |
| BC060221.1 | - | 7 - 8 | 2134 | 6843338 | 104 | |
| CT010397.1 | - | 6 - 7 | 1861 | 104 | ||
| U19463.1 | - | 6 - 7 | 1861 | 104 | ||
Set Membership: |
|