Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192726_m1
          See other KLF17 GE Assays ›
          Gene Symbol:
          Klf17
          Gene Name:
          Kruppel-like factor 17
          Gene Aliases:
          7420700M05Rik, AA420409, AU043468, C85123, D4Ertd561e, Gzf, Zfp393
          Chromosome Location:
          Chr.4: 117756659 - 117765666 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192726_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Klf17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029416.2 NP_083692.2 2 - 3 969 64
          GenBank mRNA AF508984.1 - 2 - 3 935 64
          AK018361.1 - 2 - 3 935 64
          AK139827.1 - 2 - 3 936 64
          BC064748.1 - 2 - 3 913 30476790 64

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Klf17

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          gamete generation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          nucleic acid binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription regulatory region DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 BC064748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 AK018361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 AK139827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 NM_029416.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 AF508984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 BC064748.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 AK018361.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 AK139827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 NM_029416.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P271068 AF508984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P18756 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697347 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540060 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286962 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89746 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160513 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 BC064748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 AK018361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 AK139827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 NM_029416.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 AF508984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 BC064748.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 AK018361.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 AK139827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 NM_029416.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P271068 AF508984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P18756 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 BC064748.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 AK018361.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 AK139827.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 NM_029416.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 AF508984.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 BC064748.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 AK018361.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 AK139827.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 NM_029416.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P271068 AF508984.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236374 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236387 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236382 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236384 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23236383 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160513 - -
          Whole Mouse Genome G4122A A_51_P271068 BC064748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 AK018361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 AK139827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 NM_029416.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 AF508984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 BC064748.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 AK018361.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 AK139827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 NM_029416.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P271068 AF508984.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160513 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03244088 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03858358 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03858360 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03435202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23236395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236369 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236371 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236358 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236394 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236384 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236361 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236341 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236364 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236389 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236337 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03329334 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03244087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236357 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03834270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03943396 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03943395 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236356 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236403 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03599267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236340 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04045238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23236376 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515499 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515498 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515496 - -
          Affymetrix HT MG-430 PM Array Plate 1438899_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429303_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456561_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515495 - -
          Mouse Genome 430 2.0 Array 1429303_at - -
          Mouse Genome 430 2.0 Array 1429303_PM_at - -
          Mouse Genome 430 2.0 Array 1438899_PM_at - -
          Mouse Genome 430 2.0 Array 1438899_at - -
          Mouse Genome 430 2.0 Array 1456561_PM_s_at - -
          Mouse Genome 430 2.0 Array 1456561_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75753_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75753_PM_at - -
          Mouse Expression Set 430 1429303_PM_at - -
          Mouse Expression Set 430 1429303_at - -
          Murine Genome U74v2 Set 162265_PM_r_at - -
          Murine Genome U74v2 Set 92324_PM_at - -
          Murine Genome U74v2 Set 92324_at - -
          Murine Genome U74v2 Set 162265_r_at - -
          Mouse Genome 430A 2.0 Array 1429303_at - -
          Mouse Genome 430A 2.0 Array 1429303_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429303_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1429303_at - -
          Affymetrix HT Mouse Genome 430B Array 1438899_at - -
          Affymetrix HT Mouse Genome 430B Array 1456561_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1456561_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1438899_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2320154 - -
          Illumina mouse-6 v1.0 expression beadchip 3830095 - -
          Illumina mouse-6 v1.0 expression beadchip 2320154 - -
          Illumina mouse-6 v1.0 expression beadchip 6590156 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830095 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006590156 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226178 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2469125 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227929 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227929 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2469125 - -
          mouse-6 v1.1 (Illumina) scl0075753.2_179-S - -
          mouse-6 v1.1 (Illumina) scl0075753.2_78-S - -
          mouse-6 v1.1 (Illumina) scl075753.2_67-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830095 - -
          Illumina Sentrix mouse-6.v1 bead array 6590156 - -
          Illumina Sentrix mouse-6.v1 bead array 2320154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0075753.2_179-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0075753.2_78-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl075753.2_67-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320154 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830095 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6590156 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226178 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1225165 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2469125 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1225165 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2469125 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline