Search
Search

Gtf2i| Gene Symbol: Gtf2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080746.2 | NP_001074215.1 | 18 - 19 | 2007 | 72 | |
| NM_001080747.2 | NP_001074216.1 | 17 - 18 | 1944 | 72 | ||
| NM_001080748.2 | NP_001074217.1 | 16 - 17 | 1887 | 72 | ||
| NM_001080749.2 | NP_001074218.1 | 16 - 17 | 1887 | 72 | ||
| NM_010365.4 | NP_034495.2 | 17 - 18 | 1950 | 72 | ||
| GenBank mRNA | AF017085.1 | - | 18 - 19 | 72 | ||
| AF043219.1 | - | 16 - 17 | 1571 | 72 | ||
| AF043220.1 | - | 15 - 16 | 1508 | 72 | ||
| AK145698.1 | - | 8 - 9 | 835 | 72 | ||
| AK147201.1 | - | 16 - 17 | 1845 | 72 | ||
| AY030290.1 | - | 17 - 18 | 1637 | 72 | ||
| AY030291.1 | - | 17 - 18 | 1643 | 72 | ||
| AY030292.1 | - | 16 - 17 | 1580 | 72 | ||
| AY030293.1 | - | 18 - 19 | 1700 | 72 | ||
| BC047387.1 | - | 16 - 17 | 1535 | 72 | ||
| BC053044.1 | - | 18 - 19 | 1976 | 5704935 | 72 | |
Set Membership: |
|