Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01197426_g1
          See other PLEC GE Assays ›
          Gene Symbol:
          Plec
          Gene Name:
          plectin
          Gene Aliases:
          AA591047, AU042537, EBS1, PCN, PLTN, Plec1
          Chromosome Location:
          Chr.15: 76170974 - 76231378 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01197426_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plec
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163540.1 NP_001157012.1 10 - 11 1572 83
          NM_001163542.1 NP_001157014.1 11 - 12 1089 83
          NM_001163549.1 NP_001157021.1 10 - 11 1135 83
          NM_001164203.1 NP_001157675.1 11 - 12 1076 83
          NM_011117.2 NP_035247.2 9 - 10 1038 83
          NM_201385.2 NP_958787.2 9 - 10 973 83
          NM_201386.2 NP_958788.2 9 - 10 1034 83
          NM_201387.3 NP_958789.3 9 - 10 1063 83
          NM_201388.2 NP_958790.2 9 - 10 1025 83
          NM_201389.2 NP_958791.2 9 - 10 1557 83
          NM_201390.2 NP_958792.2 9 - 10 972 83
          NM_201391.2 NP_958793.2 9 - 10 1120 83
          NM_201392.2 NP_958794.2 9 - 10 946 83
          NM_201393.2 NP_958795.2 9 - 10 1063 83
          NM_201394.2 NP_958796.2 9 - 10 1120 83
          GenBank mRNA AF188008.1 - 10 - 11 1572 83
          AF188011.1 - 10 - 11 1135 83
          AF188012.1 - 9 - 10 1120 83
          AF188013.1 - 11 - 12 1089 83
          AF188014.1 - 9 - 10 1038 83
          AF188021.1 - 9 - 10 946 83
          AF188023.1 - 11 - 12 1076 83
          AK017743.1 - 9 - 10 953 83
          AK044559.1 - 1133 83
          AY480033.1 - 9 - 10 1038 83
          AY480034.1 - 9 - 10 973 83
          AY480035.1 - 9 - 10 1034 83
          AY480036.1 - 9 - 10 1063 83
          AY480037.1 - 9 - 10 1025 83
          AY480038.1 - 9 - 10 1557 83
          AY480039.1 - 9 - 10 972 83
          AY480040.1 - 9 - 10 1120 83
          AY480041.1 - 9 - 10 946 83
          AY480042.1 - 9 - 10 1063 83
          AY480043.1 - 9 - 10 1120 83
          BC021810.1 - 9 - 10 1107 83
          BC099929.1 - 9 - 10 1558 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002346.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plec

          Panther Classification:

          Molecular Function -

          intermediate filament binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to nutrient
          hemidesmosome assembly
          actin binding
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          structural constituent of muscle
          ankyrin binding
          poly(A) RNA binding
          protein N-terminus binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001163540.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001163540.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001163542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001163542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001163549.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001163549.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001164203.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_001164203.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_011117.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_011117.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201385.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201385.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201386.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201386.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201387.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201387.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201388.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201388.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201389.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201389.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201390.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201390.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201391.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201391.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201392.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201392.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201393.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201393.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201394.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 NM_201394.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1500013461.mm.1 TC1500002346.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline