Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01198692_m1
          See other FRG1 GE Assays ›
          Gene Symbol:
          Frg1
          Gene Name:
          FSHD region gene 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 41397454 - 41417118 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01198692_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Frg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013522.3 NP_038550.2 8 - 9 896 122
          GenBank mRNA AK077107.1 - 8 - 9 823 122
          AK079229.1 - 8 - 9 788 122
          AW496369.1 - 1 - 2 122
          BC002027.1 - 8 - 9 852 3487998 122
          BE334154.1 - 1 - 2 122
          U62105.1 - 8 - 9 855 122

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002121.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Frg1

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA processing
          mRNA processing
          muscle organ development
          RNA splicing
          ribosome biogenesis
          RNA binding
          actin binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810920 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810922 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810921 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810918 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267544 U62105.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P267544 U62105.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409006 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 AK019496.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 AK028045.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 AK145992.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 AK160867.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 BC023112.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 BC060169.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 530006 NM_030165.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 911714 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 826422 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 213090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563328 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320453 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543206 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 49959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137866 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117620 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134486 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267544 U62105.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P267544 U62105.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267544 U62105.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P267544 U62105.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25810923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01720679 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117620 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06489364 - -
          Whole Mouse Genome G4122A A_51_P267544 U62105.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P267544 U62105.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135429 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117620 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01720679 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134486 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07268689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07268701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07268703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07268688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07268686 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07268697 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810920 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810921 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810918 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25810922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25810925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810893 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07034137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06744621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810894 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810899 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07268686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07018338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07268703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07248014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07018340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07270071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06701385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07157191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25810911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06865569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07019419 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10578385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578380 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578379 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578383 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578381 - -
          Affymetrix Mouse Gene 1.0 ST Array 10578382 - -
          Affymetrix HT MG-430 PM Array Plate 1417253_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1446693_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10578377 - -
          Mouse Genome 430 2.0 Array 1417253_PM_at - -
          Mouse Genome 430 2.0 Array 1417253_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14300_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14300_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800016357.mm.1 NM_013522.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016357.mm.1 NM_013522.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0800016357.mm.1 TC0800002121.mm.1 MTA TC
          Mouse Expression Set 430 1417253_PM_at - -
          Mouse Expression Set 430 1417253_at - -
          Mouse Expression Set 430 1446693_at - -
          Mouse Expression Set 430 1446693_PM_at - -
          Murine Genome U74v2 Set 100978_at - -
          Murine Genome U74v2 Set 100978_PM_at - -
          Murine Genome U74v2 Set 166522_PM_r_at - -
          Murine Genome U74v2 Set 166522_r_at - -
          Mouse Genome 430A 2.0 Array 1417253_at - -
          Mouse Genome 430A 2.0 Array 1417253_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417253_at - -
          Affymetrix HT Mouse Genome 430A Array 1417253_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1850441 - -
          Illumina mouse-6 v1.0 expression beadchip 1850441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850441 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1240867 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1240867 - -
          mouse-6 v1.1 (Illumina) scl34884.5.1_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34884.5.1_26-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1240867 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2896797 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2896797 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1240867 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline