Search
Search
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028833.3 | NP_083109.2 | 15 - 16 | 1536 | 133 | |
| GenBank mRNA | AK006472.2 | - | 14 - 15 | 1277 | 133 | |
| BC059223.1 | - | 15 - 16 | 1536 | 6417270 | 133 | |
| BC065128.1 | - | 13 - 14 | 1217 | 133 | ||
Set Membership: |
|