Search
Search

Sorcs3| Gene Symbol: Sorcs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025696.3 | NP_079972.1 | 24 - 25 | 3482 | 100 | |
| GenBank mRNA | AF276314.1 | - | 24 - 25 | 3502 | 100 | |
| AK158805.1 | - | 24 - 25 | 3349 | 100 | ||
Set Membership: |
|