| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300003973.mm.1 |
NM_001018031.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003973.mm.1 |
NM_001018031.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003973.mm.1 |
TC0300000591.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300003975.mm.1 |
NM_001018031.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003975.mm.1 |
NM_001018031.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003975.mm.1 |
TC0300000591.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300003971.mm.1 |
NM_001018031.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003971.mm.1 |
NM_001018031.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003971.mm.1 |
TC0300000591.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300003974.mm.1 |
NM_001018031.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003974.mm.1 |
NM_001018031.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300003974.mm.1 |
TC0300000591.mm.1 |
MTA TC |