Search
Search

Slc9a9| Gene Symbol: Slc9a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177909.5 | NP_808577.3 | 9 - 10 | 1264 | 91 | |
| GenBank mRNA | AK037058.1 | - | 9 - 10 | 1248 | 91 | |
| AK156494.1 | - | 9 - 10 | 1225 | 91 | ||
| AK172687.1 | - | 9 - 10 | 1236 | 91 | ||
| BC099942.1 | - | 9 - 10 | 1238 | 30103308 | 91 | |
| BC153825.1 | - | 9 - 10 | 1242 | 40093280 | 91 | |
Set Membership: |
|