Search
Search

Ednrb| Gene Symbol: Ednrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136061.2 | NP_001129533.1 | 6 - 7 | 1501 | 107 | |
| NM_001276296.1 | NP_001263225.1 | 5 - 6 | 1315 | 107 | ||
| NM_007904.4 | NP_031930.1 | 5 - 6 | 1315 | 107 | ||
| GenBank mRNA | AK076426.1 | - | 5 - 6 | 1309 | 107 | |
| AK082103.1 | - | 6 - 7 | 1302 | 107 | ||
| AK083415.1 | - | 6 - 7 | 1198 | 107 | ||
| AK085165.1 | - | 5 - 6 | 1308 | 107 | ||
| AK085532.1 | - | 5 - 6 | 1311 | 107 | ||
| BC026553.1 | - | 6 - 7 | 1427 | 4971909 | 107 | |
| U32329.1 | - | 5 - 6 | 1293 | 107 | ||
Set Membership: |
|