Search
Search
| Gene Symbol: Tut1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197993.2 | NP_932110.1 | 7 - 8 | 1479 | 62 | |
| GenBank mRNA | AK213482.1 | - | 1 - 2 | 77 | 62 | |
| BC023900.1 | - | 7 - 8 | 1456 | 5321265 | 62 | |
| BC025499.1 | - | 7 - 8 | 1503 | 5320398 | 62 | |
Set Membership: |
|