Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01232306_g1
See other FER GE Assays ›
Gene Symbol:
Fer
Gene Name:
fer (fms/fps related) protein kinase
Gene Aliases:
AV082135, C330004K01Rik, Fert, Fert2
Chromosome Location:
Chr.17: 63829950 - 64146519 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01232306_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: Fer
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_008000.2
NP_032026.2
2 - 3
290
82
GenBank mRNA
M32054.1
-
2 - 3
290
82
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the M32054.1, NM_008000.2 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Protein Kinase
Detect Genomic DNA
Mus musculus
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene Fer
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
mitotic cell cycle
regulation of protein phosphorylation
protein phosphorylation
chemotaxis
cell adhesion
signal transduction
cell proliferation
regulation of lamellipodium assembly
regulation of fibroblast migration
phosphorylation
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
cell differentiation
positive regulation of cell migration
positive regulation of actin filament polymerization
actin cytoskeleton reorganization
response to lipopolysaccharide
cellular response to insulin stimulus
negative regulation of mast cell activation involved in immune response
substrate adhesion-dependent cell spreading
cellular response to reactive oxygen species
extracellular matrix-cell signaling
cellular response to macrophage colony-stimulating factor stimulus
response to platelet-derived growth factor
insulin receptor signaling pathway via phosphatidylinositol 3-kinase
peptidyl-tyrosine autophosphorylation
Fc-epsilon receptor signaling pathway
Kit signaling pathway
regulation of epidermal growth factor receptor signaling pathway
regulation of cell proliferation
tyrosine phosphorylation of Stat3 protein
regulation of mast cell degranulation
cell-cell adhesion mediated by cadherin
innate immune response
protein autophosphorylation
platelet-derived growth factor receptor signaling pathway
diapedesis
positive regulation of NF-kappaB transcription factor activity
interleukin-6-mediated signaling pathway
nucleotide binding
actin binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
epidermal growth factor receptor binding
protein binding
ATP binding
cytoskeletal protein binding
protein phosphatase 1 binding
lipid binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
Rab GTPase binding
protein kinase binding
gamma-catenin binding
cadherin binding
cell adhesion molecule binding
MicroArray Probes:
Affymetrix
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1700010061.mm.1
NM_008000.2.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1700010061.mm.1
NM_008000.2
Refseq
Affymetrix Mouse Clariom S Array
PSR1700010061.mm.1
TC1700001053.mm.1
MTA TC
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline