Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01237815_g1
          See other APOO GE Assays ›
          Gene Symbol:
          Apoo Apoo-ps
          Gene Name:
          apolipoprotein O
          apolipoprotein O, pseudogene
          Gene Aliases:
          0610008C08Rik, 1110019O03Rik
          Apoo, EG621156
          Chromosome Location:
          Chr.X: 94367011 - 94417093 on Build GRCm38
          Chromosome Location:
          Chr.13: 107414163 - 107414767 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01237815_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199337.1 NP_001186266.1 2 - 3 458 88
          NM_001199338.1 NP_001186267.1 2 - 3 283 88
          NM_001199339.1 NP_001186268.1 2 - 3 442 88
          NM_026673.4 NP_080949.2 2 - 3 163 88
          GenBank mRNA AK002320.1 - 2 - 3 460 88
          AK003831.1 - 2 - 3 274 88
          BC016557.1 - 2 - 3 413 3489997 88
          BC107208.1 - 2 - 3 155 40051296 88
          BC107209.1 - 2 - 3 154 40051299 88
          CB595096.1 - 2 - 3 152 88
          EL957008.1 - 2 - 3 160 88
          Gene Symbol: Apoo-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_004438.1 1 149 88
          GenBank mRNA BI716015.1 - 1 118 88
          CV674789.1 - 1 134 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
          This assay is in the 5'UTR region of the AK003831.1,BC016557.1,BC107208.1,BC107209.1,NM_001199338.1,NM_001199339.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoo
          Click here to find siRNAs for Gene Apoo-ps

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0X00005737.mm.1 NM_026673.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005737.mm.1 NM_026673.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005737.mm.1 NR_004438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005737.mm.1 TC0X00000897.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 NM_001199337.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 NM_001199337.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 NM_001199338.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 NM_001199338.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 NM_001199339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 NM_001199339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005743.mm.1 TC0X00000897.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_001199337.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_001199337.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_001199338.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_001199338.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_001199339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_001199339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_026673.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NM_026673.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 NR_004438.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005747.mm.1 TC0X00000897.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 NM_001199337.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 NM_001199337.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 NM_001199338.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 NM_001199338.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 NM_001199339.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 NM_001199339.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005741.mm.1 TC0X00000897.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline