Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01238611_m1
          See other MNDAL GE Assays ›
          Gene Symbol:
          Mndal
          Gene Name:
          myeloid nuclear differentiation antigen like
          Gene Aliases:
          Gm2785
          Chromosome Location:
          Chr.1: 173857220 - 173880260 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01238611_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mndal
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170853.1 NP_001164324.1 841 102
          GenBank mRNA AK136391.1 - 843 102
          FJ594740.1 - 4 - 5 740 102
          FJ594741.1 - 749 102

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          negative regulation of cell growth
          regulation of growth
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          double-stranded DNA binding
          transcription factor binding
          identical protein binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P17218 FJ594740.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P17218 AK136391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P17218 NM_001170853.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P17218 FJ594740.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P17218 AK136391.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P17218 NM_001170853.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00151989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20839740 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1452231_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1426906_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100029547.mm.1 NM_001170853.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029547.mm.1 NM_001170853.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029547.mm.1 TC0100003890.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029548.mm.1 NM_001170853.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029548.mm.1 NM_001170853.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029548.mm.1 TC0100003890.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100031616.mm.1 NM_001170853.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031616.mm.1 NM_001170853.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100031616.mm.1 TC0100003890.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100029546.mm.1 NM_001170853.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029546.mm.1 NM_001170853.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100029546.mm.1 TC0100003890.mm.1 MTA TC
          Mouse Expression Set 430 1452348_PM_s_at - -
          Mouse Expression Set 430 1452348_s_at - -
          Murine Genome U74v2 Set 94224_PM_s_at - -
          Murine Genome U74v2 Set 94224_s_at - -
          Mouse Genome 430A 2.0 Array 1452348_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452348_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 414909 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380670 - -
          Illumina mouse-6 v1.0 expression beadchip 1980121 - -
          Illumina mouse-6 v1.0 expression beadchip 6380670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380670 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001980121 - -
          mouse-6 v1.1 (Illumina) scl000868.1_2-S - -
          mouse-6 v1.1 (Illumina) scl000854.1_75-S - -
          Illumina Sentrix mouse-6.v1 bead array 1980121 - -
          Illumina Sentrix mouse-6.v1 bead array 6380670 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000868.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000854.1_75-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1980121 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380670 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2483422 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2495068 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline