Search
Search

Adcy7| Gene Symbol: Adcy7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037723.3 | NP_001032812.2 | 17 - 18 | 2436 | 60 | |
| NM_001037724.4 | NP_001032813.1 | 17 - 18 | 2372 | 60 | ||
| NM_001109756.1 | NP_001103226.1 | 17 - 18 | 2340 | 60 | ||
| NM_007406.2 | NP_031432.2 | 17 - 18 | 2524 | 60 | ||
| GenBank mRNA | AK138911.1 | - | 17 - 18 | 2356 | 60 | |
| AK154262.1 | - | 17 - 18 | 2427 | 60 | ||
| AK154652.1 | - | 17 - 18 | 2342 | 60 | ||
| AK155646.1 | - | 17 - 18 | 2341 | 60 | ||
| AK155831.1 | - | 17 - 18 | 2343 | 60 | ||
| BC057682.1 | - | 9 - 10 | 948 | 60 | ||
| BC115833.1 | - | 15 - 16 | 2218 | 40086640 | 60 | |
| U12919.1 | - | 17 - 18 | 2525 | 60 | ||
Set Membership: |
|