Search
Search
| Gene Symbol: Sv2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022030.3 | NP_071313.1 | 11 - 12 | 2305 | 70 | |
| GenBank mRNA | AK028318.1 | - | 11 - 12 | 2250 | 70 | |
| AK080502.1 | - | 11 - 12 | 2259 | 70 | ||
| AK080525.1 | - | 11 - 12 | 2348 | 70 | ||
| AK122360.1 | - | 11 - 12 | 2341 | 70 | ||
| AK144166.1 | - | 11 - 12 | 2328 | 70 | ||
| BC026494.1 | - | 5 - 6 | 652 | 70 | ||
| BC046587.1 | - | 11 - 12 | 2305 | 6493509 | 70 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK158936.1 | - | Chr. - - [-] on Build | 70 | |||
Set Membership: |
|