Search
Search

Slc35d1| Gene Symbol: Slc35d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177732.4 | NP_808400.1 | 6 - 7 | 551 | 147 | |
| GenBank mRNA | AK049180.1 | - | 6 - 7 | 552 | 147 | |
| AK049462.1 | - | 6 - 7 | 471 | 147 | ||
| AK052968.1 | - | 6 - 7 | 471 | 147 | ||
| AK140305.1 | - | 6 - 7 | 843 | 147 | ||
| AK169775.1 | - | 6 - 7 | 453 | 147 | ||
| AK220330.1 | - | 6 - 7 | 461 | 147 | ||
| BC115984.1 | - | 7 - 8 | 452 | 40105299 | 147 | |
Set Membership: |
|