Search
Search
| Gene Symbol: Pogz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165948.1 | NP_001159420.1 | 17 - 18 | 2683 | 61 | |
| NM_172683.3 | NP_766271.2 | 17 - 18 | 2710 | 61 | ||
| GenBank mRNA | AK030880.1 | - | 5 - 6 | 606 | 61 | |
| AK035255.1 | - | 17 - 18 | 2565 | 61 | ||
| AK122288.1 | - | 7 - 8 | 867 | 61 | ||
| AK161889.1 | - | 17 - 18 | 2538 | 61 | ||
| BC032176.1 | - | 8 - 9 | 1001 | 61 | ||
| BC065134.1 | - | 17 - 18 | 2699 | 61 | ||
| BC066108.1 | - | 17 - 18 | 2699 | 61 | ||
| BC085188.1 | - | 17 - 18 | 2543 | 30682742 | 61 | |
| BC096492.1 | - | 16 - 17 | 2553 | 30745658 | 61 | |
| BC139432.1 | - | 17 - 18 | 2565 | 8862454 | 61 | |
| BC139433.1 | - | 17 - 18 | 2565 | 8862455 | 61 | |
Set Membership: |
|