Search
Search
| Gene Symbol: Wnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271678.1 | NP_001258607.1 | 20 - 21 | 4805 | 75 | |
| NM_001271679.1 | NP_001258608.1 | 20 - 21 | 4664 | 75 | ||
| GenBank mRNA | BC043119.1 | - | 3 - 4 | 858 | 75 | |
| BC060731.1 | - | 3 - 4 | 858 | 75 | ||
| JQ247189.1 | - | 20 - 21 | 4805 | 75 | ||
| JQ247190.1 | - | 20 - 21 | 4664 | 75 | ||
Set Membership: |
|