Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01271168_g1
          See other BRD2 GE Assays ›
          Gene Symbol:
          Brd2
          Gene Name:
          bromodomain containing 2
          Gene Aliases:
          AW228947, D17H6S113E, Frg-1, Fsrg-1, Fsrg1, Nat, Ring3, Rnf3, mKIAA4005
          Chromosome Location:
          Chr.17: 34112019 - 34122607 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01271168_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001204973.1 NP_001191902.1 4 - 5 2183 101
          NM_010238.3 NP_034368.2 4 - 5 2002 101
          NR_037970.1 4 - 5 1829 101
          GenBank mRNA AB010246.1 - 3 - 4 507 101
          AB010247.1 - 3 - 4 476 101
          AB010248.1 - 2 - 3 373 101
          AB212273.1 - 2 - 3 337 101
          AF045462.1 - 3 - 4 1148 101
          AF318183.1 - 3 - 4 1165 101
          AK158970.1 - 4 - 5 2003 101
          AK168423.1 - 4 - 5 1261 101
          AK168525.1 - 4 - 5 1874 101
          AK179882.1 - 3 - 4 266 101
          AK184232.1 - 140 101
          AK205877.1 - 2 - 3 174 101
          AK218127.1 - 2 - 3 174 101
          AK220444.1 - 3 - 4 1150 101
          BC046809.1 - 4 - 5 882 101
          BC085251.1 - 5 - 6 2025 101
          BC138655.1 - 3 - 4 776 8861675 101
          BC138656.1 - 3 - 4 776 8861676 101
          KJ650222.1 - 3 - 4 468 101

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brd2

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          chromatin modification
          chromatin binding
          lysine-acetylated histone binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700017739.mm.1 NR_037970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017739.mm.1 NR_037970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017739.mm.1 TC1700001898.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 NM_001204973.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 NM_001204973.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 NM_010238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 NM_010238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 NR_037970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 NR_037970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017738.mm.1 TC1700001898.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 NM_001204973.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 NM_001204973.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 NM_010238.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 NM_010238.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 NR_037970.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 NR_037970.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017737.mm.1 TC1700001898.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline