Search
Search

Alppl2| Gene Symbol: Alppl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007433.3 | NP_031459.3 | 9 - 10 | 1347 | 102 | |
| GenBank mRNA | AK145571.1 | - | 10 - 11 | 1256 | 102 | |
| AY054302.1 | - | 9 - 10 | 1188 | 102 | ||
| BC052874.1 | - | 9 - 10 | 1342 | 30033693 | 102 | |
Set Membership: |
|