Search
Search
| Gene Symbol: Plp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290561.1 | NP_001277490.1 | 4 - 5 | 687 | 85 | |
| NM_011123.3 | NP_035253.1 | 4 - 5 | 792 | 85 | ||
| GenBank mRNA | AK134554.1 | - | 4 - 5 | 783 | 85 | |
| AK160872.1 | - | 4 - 5 | 787 | 85 | ||
| AK197253.1 | - | 1 - 2 | 173 | 85 | ||
| AK213737.1 | - | 1 - 2 | 173 | 85 | ||
| AK215067.1 | - | 1 - 2 | 173 | 85 | ||
| BC027010.1 | - | 4 - 5 | 702 | 5364736 | 85 | |
| M15442.1 | - | 4 - 5 | 771 | 85 | ||
Set Membership: |
|