Search
Search

Ddx17| Gene Symbol: Ddx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040187.1 | NP_001035277.1 | 5 - 6 | 1002 | 66 | |
| NM_199079.1 | NP_951061.1 | 5 - 6 | 821 | 66 | ||
| NM_199080.2 | NP_951062.1 | 5 - 6 | 1002 | 66 | ||
| GenBank mRNA | AK039888.1 | - | 5 - 6 | 821 | 66 | |
| AK151061.1 | - | 5 - 6 | 777 | 66 | ||
| AK152749.1 | - | 5 - 6 | 777 | 66 | ||
| AK152831.1 | - | 5 - 6 | 777 | 66 | ||
| AK161001.1 | - | 5 - 6 | 818 | 66 | ||
| AK161012.1 | - | 5 - 6 | 817 | 66 | ||
| BC062910.1 | - | 5 - 6 | 807 | 6816663 | 66 | |
| BC096036.1 | - | 5 - 6 | 1002 | 30549388 | 66 | |
Set Membership: |
|