Search
Search

Haghl| Gene Symbol: Haghl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271433.1 | NP_001258362.1 | 2 - 3 | 350 | 64 | |
| NM_001271434.1 | NP_001258363.1 | 2 - 3 | 350 | 64 | ||
| NM_001271435.1 | NP_001258364.1 | 2 - 3 | 350 | 64 | ||
| NM_026897.3 | NP_081173.1 | 1 - 2 | 335 | 64 | ||
| GenBank mRNA | AK005274.1 | - | 1 - 2 | 331 | 64 | |
| AK012748.1 | - | 2 - 3 | 347 | 64 | ||
| AK034798.1 | - | 2 - 3 | 325 | 64 | ||
| AK047517.1 | - | 2 - 3 | 305 | 64 | ||
| AK079632.1 | - | 1 - 2 | 333 | 64 | ||
| BC005586.1 | - | 2 - 3 | 296 | 64 | ||
| BC030466.1 | - | 2 - 3 | 335 | 5253213 | 64 | |
| BC083322.1 | - | 2 - 3 | 289 | 6820885 | 64 | |
Set Membership: |
|