Search
Search

Dhx58| Gene Symbol: Dhx58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030150.2 | NP_084426.2 | 4 - 5 | 668 | 77 | |
| GenBank mRNA | AF316999.1 | - | 3 - 4 | 567 | 77 | |
| AK153342.1 | - | 4 - 5 | 597 | 77 | ||
| BC029209.1 | - | 3 - 4 | 2651254 | 77 | ||
Set Membership: |
|