Search
Search

Dazap2| Gene Symbol: Dazap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011873.2 | NP_036003.2 | 2 - 3 | 181 | 72 | |
| GenBank mRNA | AF085348.1 | - | 2 - 3 | 146 | 72 | |
| AK002595.1 | - | 2 - 3 | 218 | 72 | ||
| AK050159.1 | - | 2 - 3 | 195 | 72 | ||
| AK088916.1 | - | 2 - 3 | 203 | 72 | ||
| AK132139.1 | - | 2 - 3 | 195 | 72 | ||
| AK137246.1 | - | 2 - 3 | 182 | 72 | ||
| AK145398.1 | - | 2 - 3 | 215 | 72 | ||
| AK146970.1 | - | 2 - 3 | 206 | 72 | ||
| BC014808.1 | - | 2 - 3 | 181 | 4506124 | 72 | |
| BC029351.1 | - | 1 - 2 | 112 | 72 | ||
| BC083347.1 | - | 2 - 3 | 173 | 6829350 | 72 | |
Set Membership: |
|