Search
Search

Gmpr2| Gene Symbol: Gmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177992.2 | NP_818773.1 | 2 - 3 | 244 | 66 | |
| GenBank mRNA | AK017821.1 | - | 2 - 3 | 539 | 66 | |
| AK031004.1 | - | 2 - 3 | 246 | 66 | ||
| AK075772.1 | - | 2 - 3 | 202 | 66 | ||
| BC003886.1 | - | 2 - 3 | 203 | 3590411 | 66 | |
| BC024109.1 | - | 2 - 3 | 200 | 5149353 | 66 | |
| BY099107.1 | - | 2 - 3 | 245 | 66 | ||
Set Membership: |
|