Search
Search
| Gene Symbol: Ostc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025509.3 | NP_079785.1 | 2 - 3 | 302 | 67 | |
| GenBank mRNA | AK011085.1 | - | 2 - 3 | 254 | 67 | |
| AK013011.1 | - | 2 - 3 | 261 | 67 | ||
| AK135332.1 | - | 2 - 3 | 258 | 67 | ||
| BC021935.1 | - | 2 - 3 | 276 | 5149713 | 67 | |
| BY228396.1 | - | 2 - 3 | 304 | 67 | ||
Set Membership: |
|