Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01331557_m1
          See other AMBRA1 GE Assays ›
          Gene Symbol:
          Ambra1
          Gene Name:
          autophagy/beclin 1 regulator 1
          Gene Aliases:
          2310079H06Rik, A130023A14, AA474864, AV021921, D030051N19Rik, mKIAA1736
          Chromosome Location:
          Chr.2: 91730111 - 91918849 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01331557_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ambra1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080754.1 NP_001074223.1 8 - 9 2169 70
          NM_172669.3 NP_766257.3 7 - 8 2442 70
          GenBank mRNA AK083596.1 - 7 - 8 2399 70
          AK084557.1 - 7 - 8 2424 70
          AK129433.1 - 3 - 4 1706 70
          AK155513.1 - 7 - 8 2405 70
          AK169280.1 - 8 - 9 2122 70
          BC032293.1 - 8 - 9 2092 70
          BC054082.1 - 7 - 8 2374 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001382.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ambra1

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagy
          multicellular organism development
          nervous system development
          negative regulation of cell proliferation
          embryo development
          neural tube development
          cell differentiation
          negative regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          mitophagy in response to mitochondrial depolarization
          positive regulation of mitophagy
          regulation of autophagosome assembly
          protein binding
          ubiquitin protein ligase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200010884.mm.1 NM_001080754.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010884.mm.1 NM_001080754.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010884.mm.1 NM_172669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010884.mm.1 NM_172669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010884.mm.1 TC0200001382.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200010883.mm.1 NM_001080754.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010883.mm.1 NM_001080754.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010883.mm.1 NM_172669.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010883.mm.1 NM_172669.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200010883.mm.1 TC0200001382.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline