Search
Search
| Gene Symbol: Hk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033245.4 | NP_001028417.1 | 7 - 8 | 988 | 58 | |
| NM_001206390.1 | NP_001193319.1 | 7 - 8 | 798 | 58 | ||
| NM_001206391.1 | NP_001193320.1 | 7 - 8 | 798 | 58 | ||
| NM_001206392.1 | NP_001193321.1 | 6 - 7 | 868 | 58 | ||
| GenBank mRNA | AK140444.1 | - | 7 - 8 | 852 | 58 | |
| AK149651.1 | - | 7 - 8 | 838 | 58 | ||
| AK149992.1 | - | 7 - 8 | 800 | 58 | ||
| AK152839.1 | - | 1 - 2 | 92 | 58 | ||
| AK154603.1 | - | 7 - 8 | 960 | 58 | ||
| AK155701.1 | - | 7 - 8 | 845 | 58 | ||
| AK159857.1 | - | 6 - 7 | 714 | 58 | ||
| AK159906.1 | - | 7 - 8 | 898 | 58 | ||
| AK160018.1 | - | 7 - 8 | 898 | 58 | ||
| AK160063.1 | - | 7 - 8 | 898 | 58 | ||
| AK162637.1 | - | 7 - 8 | 931 | 58 | ||
| AK163658.1 | - | 7 - 8 | 852 | 58 | ||
| AK170813.1 | - | 6 - 7 | 58 | |||
| AK171214.1 | - | 7 - 8 | 926 | 58 | ||
| AK171248.1 | - | 7 - 8 | 838 | 58 | ||
| AK171580.1 | - | 7 - 8 | 938 | 58 | ||
| BC028480.1 | - | 7 - 8 | 801 | 58 | ||
| BC117860.1 | - | 7 - 8 | 883 | 40095265 | 58 | |
| BC117861.1 | - | 7 - 8 | 883 | 40095267 | 58 | |
Set Membership: |
|