Search
Search

Shroom2| Gene Symbol: Shroom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290684.1 | NP_001277613.1 | 3 - 4 | 485 | 86 | |
| NM_172441.3 | NP_766029.2 | 3 - 4 | 485 | 86 | ||
| GenBank mRNA | BF463332.1 | - | 3 - 4 | 322 | 86 | |
| DQ358971.1 | - | 3 - 4 | 466 | 86 | ||
| EF071946.1 | - | 3 - 4 | 485 | 86 | ||
Set Membership: |
|