Search
Search

Umodl1| Gene Symbol: Umodl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177465.4 | NP_803416.2 | 18 - 19 | 3695 | 62 | |
| GenBank mRNA | AK040446.1 | - | 10 - 11 | 2196 | 62 | |
| AY771616.1 | - | 19 - 20 | 3612 | 62 | ||
| AY771617.1 | - | 18 - 19 | 3699 | 62 | ||
| AY771618.1 | - | 18 - 19 | 3354 | 62 | ||
| BC051427.1 | - | 18 - 19 | 3350 | 62 | ||
| BC132381.1 | - | 5 - 6 | 884 | 62 | ||
Set Membership: |
|