Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01704846_s1
          See other IFIT3 GE Assays ›
          Gene Symbol:
          Ifit3
          Gene Name:
          interferon-induced protein with tetratricopeptide repeats 3
          Gene Aliases:
          Ifi49, P49
          Chromosome Location:
          Chr.19: 34583142 - 34588982 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01704846_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ifit3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          RefSeq NM_010501.2 NP_034631.1 2 1645 115
          GenBank mRNA AK077459.1 - 2 1644 115
          AK151032.1 - 2 1645 115
          AK153226.1 - 2 1646 115
          BC003804.1 - 2 1611 115
          BC089563.1 - 2 1643 115
          L32974.1 - 2 1626 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ifit3

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          cellular response to interferon-alpha
          cellular response to interferon-beta
          innate immune response
          defense response to virus
          protein binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 BC003804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 BC089563.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 L32974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 AK077459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 AK151032.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 BC003804.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 BC089563.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 L32974.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 AK077459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P359570 AK151032.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 BC003804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 BC089563.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 L32974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 AK077459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 AK151032.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 BC003804.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 BC089563.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 L32974.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 AK077459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P359570 AK151032.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32081029 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09158478 - -
          Whole Mouse Genome G4122A A_51_P359570 BC003804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 BC089563.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 L32974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 AK077459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 AK151032.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 BC003804.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 BC089563.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 L32974.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 AK077459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P359570 AK151032.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128030 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32081104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14317828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14490460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14252884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14462352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081076 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081030 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14490458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32081021 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10462619 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462620 - -
          Affymetrix HT MG-430 PM Array Plate 1449025_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10462618 - -
          Mouse Genome 430 2.0 Array 1435529_PM_at - -
          Mouse Genome 430 2.0 Array 1449025_PM_at - -
          Mouse Genome 430 2.0 Array 1449025_at - -
          Mouse Genome 430 2.0 Array 1435529_at - -
          Affymetrix Mouse Clariom S Array PSR1900004464.mm.1 NM_010501.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004464.mm.1 NM_010501.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004464.mm.1 TC1900000501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004469.mm.1 NM_010501.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004469.mm.1 NM_010501.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004469.mm.1 TC1900000501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004465.mm.1 NM_010501.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004465.mm.1 NM_010501.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004465.mm.1 TC1900000501.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900004466.mm.1 NM_010501.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004466.mm.1 NM_010501.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900004466.mm.1 TC1900000501.mm.1 MTA TC
          Mouse Expression Set 430 1449025_PM_at - -
          Mouse Expression Set 430 1449025_at - -
          Murine Genome U74v2 Set 93956_PM_at - -
          Murine Genome U74v2 Set 93956_at - -
          Mouse Genome 430A 2.0 Array 1449025_PM_at - -
          Mouse Genome 430A 2.0 Array 1449025_at - -
          Affymetrix HT Mouse Genome 430A Array 1449025_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449025_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2450278 - -
          MouseWG-6 v2 Expression BeadChip 6200113 - -
          Illumina mouse-6 v1.0 expression beadchip 3190113 - -
          Illumina mouse-6 v1.0 expression beadchip 2450278 - -
          Illumina mouse-6 v1.0 expression beadchip 6200113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003190113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006200113 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230458 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2699233 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230458 - -
          mouse-6 v1.1 (Illumina) scl0015959.1_2-S - -
          mouse-6 v1.1 (Illumina) scl015959.2_27-S - -
          mouse-6 v1.1 (Illumina) scl015959.2_49-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450278 - -
          Illumina Sentrix mouse-6.v1 bead array 3190113 - -
          Illumina Sentrix mouse-6.v1 bead array 6200113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015959.2_27-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0015959.1_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015959.2_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3190113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6200113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2944666 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246194 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230458 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230458 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246194 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2944666 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0015959.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl015959.2_49-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl015959.2_27-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch