Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02345140_g1
          See other SPAG8 GE Assays ›
          Gene Symbol:
          Spag8
          Gene Name:
          sperm associated antigen 8
          Gene Aliases:
          MH-SPAG8
          Chromosome Location:
          Chr.4: 43651330 - 43654124 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02345140_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spag8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007463.1 NP_001007464.1 5 - 6 1169 129
          NM_001290462.1 NP_001277391.1 5 - 6 1169 129
          GenBank mRNA AK132968.1 - 5 - 6 1170 129
          AY769082.1 - 5 - 6 1169 129
          BC150868.1 - 5 - 6 1211 9087779 129

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spag8

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein binding
          positive regulation of transcription from RNA polymerase II promoter
          protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115187 AY769082.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115187 NM_001007463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115187 AK132968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P115187 BC150868.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 134718 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115187 AY769082.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115187 NM_001007463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115187 AK132968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P115187 BC150868.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22873000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872979 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872981 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22872997 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00858738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03547844 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03547821 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03547843 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03547832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22873023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22872989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872944 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03431959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03547841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22872942 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10512605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10512600 - -
          Affymetrix HT MG-430 PM Array Plate 1435997_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10512598 - -
          Mouse Genome 430 2.0 Array 1435997_PM_at - -
          Mouse Genome 430 2.0 Array 1435997_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433700_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 433700_at - -
          Mouse Expression Set 430 1435997_PM_at - -
          Mouse Expression Set 430 1435997_at - -
          Affymetrix HT Mouse Genome 430B Array 1435997_at - -
          Affymetrix HT Mouse Genome 430B Array 1435997_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160526 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160526 - -
          mouse-6 v1.1 (Illumina) GI_55925584-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160526 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline