Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm02619580_g1
          See other ACTB GE Assays ›
          Gene Symbol:
          Actb
          Gene Name:
          actin, beta
          Gene Aliases:
          Actx, E430023M04Rik, beta-actin
          Chromosome Location:
          Chr.5: 142903116 - 142906724 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm02619580_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
          GenBank mRNA AK075973.1 - 3 341 143
          AK078935.1 - 3 343 143
          AK088691.1 - 3 340 143
          AK145191.1 - 3 343 143
          AK145196.1 - 3 343 143
          AK145308.1 - 3 343 143
          AK147787.1 - 3 340 143
          AK149920.1 - 3 343 143
          AK150545.1 - 3 344 143
          AK150553.1 - 3 344 143
          AK150558.1 - 3 344 143
          AK150570.1 - 3 345 143
          AK150662.1 - 3 342 143
          AK150675.1 - 3 342 143
          AK150711.1 - 3 343 143
          AK150879.1 - 3 343 143
          AK151010.1 - 3 340 143
          AK151136.1 - 3 343 143
          AK151145.1 - 3 340 143
          AK151159.1 - 3 343 143
          AK151166.1 - 3 340 143
          AK151190.1 - 3 343 143
          AK151202.1 - 3 343 143
          AK151226.1 - 3 340 143
          AK151277.1 - 3 340 143
          AK151297.1 - 3 343 143
          AK151350.1 - 3 340 143
          AK151385.1 - 3 343 143
          AK151386.1 - 3 340 143
          AK151398.1 - 3 343 143
          AK151444.1 - 3 343 143
          AK151468.1 - 3 340 143
          AK151663.1 - 3 343 143
          AK151712.1 - 3 344 143
          AK151898.1 - 3 341 143
          AK151956.1 - 3 340 143
          AK151995.1 - 3 343 143
          AK151999.1 - 3 343 143
          AK152043.1 - 3 344 143
          AK152052.1 - 3 341 143
          AK152083.1 - 3 344 143
          AK152218.1 - 3 344 143
          AK152433.1 - 3 341 143
          AK152470.1 - 3 341 143
          AK152591.1 - 3 344 143
          AK152615.1 - 3 343 143
          AK152651.1 - 3 343 143
          AK152671.1 - 3 344 143
          AK152813.1 - 3 344 143
          AK152841.1 - 3 344 143
          AK152844.1 - 3 340 143
          AK153461.1 - 3 340 143
          AK159759.1 - 3 343 143
          AK159834.1 - 3 340 143
          AK160029.1 - 3 340 143
          AK166349.1 - 3 343 143
          AK166498.1 - 3 343 143
          AK167117.1 - 3 343 143
          AK167825.1 - 3 343 143
          AK167960.1 - 3 343 143
          AK202772.1 - 1 154 143
          BC138611.1 - 3 308 143
          BC138614.1 - 3 308 143
          EF095208.1 - 2 232 143
          J04181.1 - 3 324 143
          M12481.1 - 2 184 143
          X03672.1 - 3 342 143
          X03765.1 - 2 184 143

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Endogenous Controls Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch