Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03052613_s1
          See other CYP2B13 GE Assays ›
          Gene Symbol:
          Cyp2b13
          Gene Name:
          cytochrome P450, family 2, subfamily b, polypeptide 13
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 26061344 - 26096197 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03052613_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp2b13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007813.2 NP_031839.2 9 1749 145

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp2b13

          Panther Classification:

          Molecular Function -

          oxidoreductase oxygenase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          epoxygenase P450 pathway
          exogenous drug catabolic process
          oxidation-reduction process
          arachidonic acid epoxygenase activity
          steroid hydroxylase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          oxygen binding
          heme binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P289091 BC145895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P289091 NM_007813.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P289091 BC145895.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P289091 NM_007813.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451981 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336257 AF290198.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336257 NM_148949.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336257 XM_001472405.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 336257 XM_975400.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651935 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 257402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 143173 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P289091 BC145895.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P289091 NM_007813.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P289091 BC145895.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P289091 NM_007813.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492339 BC145895.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P492339 NM_007813.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P289091 BC145895.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P289091 NM_007813.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P289091 BC145895.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P289091 NM_007813.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012569 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25012607 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144904 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01487649 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144856 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140292 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01487624 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144831 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01487633 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144757 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01487650 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06144928 - -
          Whole Mouse Genome G4122A A_52_P289091 BC145895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289091 NM_007813.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289091 BC145895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P289091 NM_007813.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492339 BC145895.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492339 NM_007813.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132470 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01487650 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06144904 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06144831 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06144856 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06144790 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25012942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25013173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05931832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25013168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05904441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25012959 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551210 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551212 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551211 - -
          Affymetrix HT MG-430 PM Array Plate 1449479_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551209 - -
          Mouse Genome 430 2.0 Array 1449479_PM_at - -
          Mouse Genome 430 2.0 Array 1449479_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13089_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13089_at - -
          Affymetrix Mouse Clariom S Array PSR0700003361.mm.1 TC0700000433.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003361.mm.1 NM_007813.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003361.mm.1 NM_007813.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003360.mm.1 TC0700000433.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003360.mm.1 NM_007813.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003360.mm.1 NM_007813.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003359.mm.1 TC0700000433.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700003359.mm.1 NM_007813.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700003359.mm.1 NM_007813.2 Refseq
          Mouse Expression Set 430 1449479_PM_at - -
          Mouse Expression Set 430 1449479_at - -
          Murine Genome U74v2 Set 102820_at - -
          Murine Genome U74v2 Set 102820_PM_at - -
          Mouse Genome 430A 2.0 Array 1449479_at - -
          Mouse Genome 430A 2.0 Array 1449479_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449479_at - -
          Affymetrix HT Mouse Genome 430A Array 1449479_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214531 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1214531 - -
          mouse-6 v1.1 (Illumina) scl013089.9_98-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013089.9_98-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1214531 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214531 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0